- Description:HSM 2.0 Instrument
- Cat. No.:PAA2715
- Supplier no.:A2715
The Heater Shaker Magnet Instrument (HSM 2.0) performs all the functions necessary for the processing of magnetic resin-based purification chemistries in large-volume formats.
- All-in-one processing for large-volume purification chemistries
- Processes magnetic resin-based purification chemistries in large-volume formats
- Uses standard 50ml conical tubes, magnets and reagent-based paramagnetic particles
- Preprogrammed with methods for processing up to 32 whole blood samples
The Heater Shaker Magnet Instrument (HSM 2.0) is designed to perform all of the functions necessary for the processing of magnetic resin-based purification chemistries in large-volume formats. With its ability to heat, shake and apply a magnetic field, the HSM 2.0 Instrument provides all-in-one processing capabilities for a variety of large-volume purification chemistries in either a manual or automated format. The instrument uses standard 50ml conical tubes, magnets and reagent-based paramagnetic particles (PMPs). The PMPs provide a mobile solid phase that optimizes capture, washing and elution of biological target molecules. Initially designed to run the ReliaPrep Large Volume HT gDNA Isolation System (Cat.# A2751), the HSM 2.0 Instrument is supplied with software containing preprogrammed isolation methods for processing up to 32 samples of human whole blood in approximately 2-3.5 hours, depending on sample volume and number. Samples are processed in a semi-automated method with the user dispensing and aspirating reagents from the samples as directed by the software on a computer screen. The programmed methods control the heating, shaking, magnetization and timing of the steps required for the semi-automated purification. For fully automated purification, the HSM 2.0 Instrument can be integrated with a robotic liquid-handling workstation, which can process 32 samples in less than 4.5 hours. Minimum Software Computer Requirements: Windows PC; Dual-Core x86-based processor, 2MB Memory, 100GB HD, Video 1024 x 768; Microsoft Windows 7 Professional and Ultimate editions (32-bit or 64-bit). Use of up-to-date antivirus software is strongly recommended.