ContactUsLinkComponent
E-A-R™ UltraFit™ 27 Earplugs, 3M™
E-A-R™ UltraFit™ 27 Earplugs, 3M™
Catalog # 89107-442
Supplier:  3M
CSA Certified
E-A-R™ UltraFit™ 27 Earplugs, 3M™
Catalog # 89107-442
Supplier:  3M
Supplier Number:  7000127188
CSA Certified
Specifications
  • Noise reduction rating:
    27 dB
  • Color:
    Yellow
  • Description:
    UltraFit™ 27 multi-flange PVC corded earplugs
  • Size:
    Universal
  • Cat. No.:
    89107-442
  • Packaging:
    Poly bag
  • Supplier no.:
    340-8002
Specifications
About this item
3M™ E-A-R™ UltraFit™ corded earplugs feature a proprietary pre-molded, triple-flange design that fits comfortably in most ear canals.
  • Ergonomic pistol grip for an easier fit
  • Triple-flange provides proven protection
  • Conforms well to the ear's natural oval shape
  • Washable and reusable
  • Noise Reduction Rating (NRR): 27 dB
Polymer formulation gently hugs the ear canal to provide an effective seal that protects against noise.
Certifications: CSA Class AL