Order Entry
United States
ContactUsLinkComponent
Fetal Bovine Serum, U.S. Origin, Rockland™
Fetal Bovine Serum, U.S. Origin, Rockland™
Catalog # 10802-768
Fetal Bovine Serum, U.S. Origin, Rockland™
Catalog # 10802-768
Supplier Number:  FBS-02-0500
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Fetal Bovine Serum, U.S. Origin
  • Size:
    500 mL
  • Environmentally Preferable:
  • Endotoxin:
    <1.0 EU/mL
  • Cat. No.:
    10802-768
  • Supplier no.:
    FBS-02-0500

Specifications

About this item

Fetal bovine serum is used in a wide range of cell culture apllications.

  • Sterile bacteriphage
  • Store at −20 °C

This product originates in the United States and is mycoplasma, virus, performance and endotoxin tested.

Caution: For in vitro diagnostic use only, not for human or animal therapeutic use.