Order Entry
United States
ContactUsLinkComponent
 
Parteck® Delta M 98.0-101.0% (by HPLC on dried basis), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, E421, SAFC®
Catalog # EM1.12635.9025
Supplier:  MilliporeSigma
CAS Number:  
undefined
Parteck® Delta M 98.0-101.0% (by HPLC on dried basis), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, E421, SAFC®
Catalog # EM1.12635.9025
Supplier:  MilliporeSigma
Supplier Number:  1.12635.9025
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Fibre carton
  • Size:
    25 kg
  • Cat. No.:
    EM1.12635.9025
  • Supplier no.:
    1.12635.9025

Specifications

Specification Test Results

  • Assay (HPLC, calc. on dried substance):
    98.0 - 101.0 %
  • Identity (NIR):
    passes test
  • Appearance of solution (10 %; water):
    passes test
  • pH-value (10 %; water):
    4.5 - 7.5
  • Melting point:
    165 - 169
  • Conductivity (25°C) (20 %; water):
    ≤ 20
  • Heavy metals (as Pb):
    ≤ 5 ppm
  • Ni (Nickel):
    ≤ 1 ppm
  • Pb (Lead):
    ≤ 0.5 ppm
  • Ru (Ruthenium) (#):
    ≤ 1 ppm
  • Related substances (HPLC) (Sorbitol (Impurity A)):
    ≤ 2.0 %
  • Related substances (HPLC) (Sum of Maltitol and Isomalt (Impurities B and C)):
    ≤ 2.0
  • Related substances (HPLC) (Major unspecified impurity):
    ≤ 0.10 %
  • Related substances (HPLC) (Sum of all impurities):
    ≤ 2.0 %
  • Reducing sugars (as glucose):
    ≤ 0.05
  • Reducing sugars after hydrolysis/total sugar (as glucose):
    ≤ 1
  • Sulfated ash:
    ≤ 0.1
  • Water:
    ≤ 0.5
  • Loss on Drying (105 °C):
    ≤ 0.30 %
  • Residual solvents (ICH Q3C):
    excluded by manufacturing process
  • Microbiological test:
    passes test
  • Colony count (aerobic bacteria (TAMC)):
    ≤ 10³
  • Colony count (Yeasts and moulds (TYMC)):
    ≤ 10²
  • Salmonella spp.:
    in 10g Substance not detectable
  • E.coli:
    in 1g Substance not detectable
  • Staphylococcus aureus:
    in 1g Substance not detectable
  • Pseudomonas aeruginosa:
    in 1g Substance not detectable
  • Candida albicans:
    In 1g Substance not detectable

Specification Test Results

About this item

 :
 :182.17 g/mol
Boiling Pt:290…295 °C
Melting Pt:164…169 °C
Storage Temperature:Ambient
EINECS:200-711-8
CAS Number:69-65-8
Formula:HOH₂C(CH(OH))₄CH₂OH
MDL Number:MFCD00064287