AttoPhos Buffer, Promega
Catalog # PAS1021
Supplier: Promega Corporation
CAS Number:
Specifications
- Description:AttoPhos Buffer
- Size:60 mL
- Storage Temperature:2 °C to 10 °C
- Cat. No.:PAS1021
- Supplier no.:S1021
Specifications
About this item
The AttoPhos(R) Buffer is optimized for sensitive detection of alkaline phosphatase (AP) when used to prepare working solutions with AttoPhos(R) Substrate (Cat.# S1011, S1012, S1013).