Lysis Buffer, Promega
Catalog # PAMD1392
Supplier: Promega Corporation
CAS Number:
Specifications
- Description:Lysis Buffer
- Size:160 mL
- Storage Temperature:15°C to 30°C
- Cat. No.:PAMD1392
- Supplier no.:MD1392
Specifications
About this item
Lysis Buffer, Blood, is a component of the MagneSil Blood Genomic, Max Yield System (Cat.# MD1360), and MagneSil ONE, Fixed Yield Blood Genomic System (Cat.# MD1370).
- Lysis Buffer Used With Automated Blood DNA Purification Systems
- Component of the MagneSil Blood Genomic, Max Yield System (Cat.# MD1360), and MagneSil ONE, Fixed Yield Blood Genomic System (Cat.# MD1370).
Lysis Buffer, Blood, is a component of the MagneSil Blood Genomic, Max Yield System (Cat.# MD1360), and MagneSil ONE, Fixed Yield Blood Genomic System (Cat.# MD1370).