Order Entry
United States
Orders LinkContactUsLinkComponent
Lysis Buffer A, Promega

Lysis Buffer A, Promega

Lysis Buffer A, 100 mL
Catalog # PAA8191
Supplier:   Promega Corporation
Supplier #:  A8191
Lysis Buffer A, Promega
Catalog # PAA8191
CAS Number:  
Lysis Buffer A, Promega
Lysis Buffer A, Promega

Lysis Buffer A, 100 mL

CAS Number:  
Price
$148.62
Log in for availability
Each (100ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Components of the Wizard Magnetic DNA Purification System for Food, which is intended for isolating genomic DNA from a variety of food samples including: corn seeds, corn meal, soy beans, soy flour, soy milk, corn oil, soybean oil, canola oil, etc.

  • Lysis Buffers Used in Protocol for Isolating DNA from Food
  • Components of the Wizard(R) Magnetic DNA Purification System for Food available as standalone buffers

Lysis Buffer A, Food, and Lysis Buffer B, Food, are comonents of the Wizard Magnetic DNA Purification System for Food. The system is intended for isolating genomic DNA from a variety of food samples including: corn seeds, corn meal, soy beans, soy flour, soy milk, corn oil, soybean oil, canola oil, etc. The DNA extracted from the food samples is suitable for use in polymerase chain reaction (PCR) testing for Genetically Modified Organism (GMO) DNA sequences.

Documents

Specifications

Specifications

  • Description:
    Lysis Buffer A
  • Size:
    100 mL
  • Cat. No.:
    PAA8191
  • Storage Temperature:
    15°C to 30°C

Specifications