Order Entry
United States
ContactUsLinkComponent
EDTA 10 mM, (pH 8.0), Promega
EDTA 10 mM, (pH 8.0), Promega
Catalog # PAA2631
CAS Number:  
undefined
EDTA 10 mM, (pH 8.0), Promega
Catalog # PAA2631
Supplier Number:  A2631
CAS Number:  

Specifications

  • Description:
    EDTA 10mM, (pH 8.0)
  • Size:
    10 mL
  • Storage Temperature:
    15°C to 30°C
  • Cat. No.:
    PAA2631
  • Supplier no.:
    A2631

Specifications

About this item

EDTA is a chelator of divalent cations suitable for biochemistry and molecular biology applications.

  • Available in 0.5M or 10mM concentrations
  • 10mM concentration also provided as part of Maxwell(R) HT 96 gDNA Blood Isolation System (Cat.# A2670, A2671)