Order Entry
United States
ContactUsLinkComponent
Calcium chloride dihydrate ≥99% (by complexometric analysis), white solid, Molecular Biology Grade for molecular biology, Calbiochem®
Calcium chloride dihydrate ≥99% (by complexometric analysis), white solid, Molecular Biology Grade for molecular biology, Calbiochem®
Catalog # 80501-994
Supplier:  MilliporeSigma
CAS Number:  
Calcium chloride dihydrate ≥99% (by complexometric analysis), white solid, Molecular Biology Grade for molecular biology, Calbiochem®
Catalog # 80501-994
Supplier:  MilliporeSigma
Supplier Number:  208291-250GM
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Size:
    250 g
  • Environmentally Preferable:
  • Cat. No.:
    80501-994
  • Supplier no.:
    208291-250GM

Specifications

Specification Test Results

  • Purity (by complexometry):
    ≥99%
  • Heavy Metals:
    ≤0.0005% (as Pb)

Specification Test Results

About this item

Synonyms: Calcium Chloride Dihydrate, Calcium dichloride
 :
 :147.01 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:10035-04-8
Formula:CaCl₂·2H₂O
MDL Number:MFCD00149613