


Manganese(II) chloride tetrahydrate ≥99%, Puriss. p.a. ACS, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:13446-34-9
:
Formula:MnCl₂·4H₂O
MDL Number:MFCD00149792
:197.9 g/mol
Boiling Pt: Dec. (on heating)
Melting Pt:58 °C
Density:1.913 g/cm³
UN:3288
ADR:6.1,III
:GHS05
:GHS08
:GHS06
Storage Temperature:Ambient
Specification Test Results
- Assay:99.0 - 101.0 %
- pH (5 %, 25 °C):3.5 - 6.0
- Calcium (Ca):max. 0.001 %
- Copper (Cu):max. 0.0005 %
- Iron (Fe):max. 5 ppm
- Potassium (K):max. 0.001 %
- Lead (Pb):max. 0.0005 %
- Magnesium (Mg):Max. 0.005 %
- Sulfate (SO₄):max. 0.005 %
- Sodium (Na):Max. 0.005 %
- Nickel (Ni):Max. 0.0005 %
- Zinc (Zn):Max. 0.001 %
- Heavy metals (ICP) ACS:Max. 5 ppm
- KMnO₄ reducing matter (as O):Max. 0.0005 %
- Water insoluble matter:Max. 0.005 %
Specification Test Results
Frequently Bought Together