J&W DB-FFAP GC Columns, Agilent Technologies
Supplier: Avantor
Certificates
About this item
DB-FFAP is designed for the analysis of volatile fatty acids and phenols.
- Bonded and cross-linked; solvent rinsable
- Close equivalent to USP Phase
- High sensitivity and efficiency
Agilent JW DB-FFAP is a nitroterephthalic-acid-modified polyethylene glycol (PEG) column of high polarity.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.