ContactUsLinkComponent
Versaflo™ Easy Clean Powered Air Purifying Respirator (PAPR) Kit TR-600-ECK, 3M™
Versaflo™ Easy Clean Powered Air Purifying Respirator (PAPR) Kit TR-600-ECK, 3M™
Catalog # 10152-830
Supplier:  3M
Versaflo™ Easy Clean Powered Air Purifying Respirator (PAPR) Kit TR-600-ECK, 3M™
Catalog # 10152-830
Supplier:  3M
Supplier Number:  7100035754
Specifications
  • Description:
    Belt mounted PARP kit, NIOSH approved
  • Battery Type:
    Standard capacity lithium ion, rechargeable
  • Headtop:
    Hood with suspension
  • Cat. No.:
    10152-830
  • Supplier no.:
    00051131373754
Specifications
About this item
Perfect for many pharmaceutical, food safety, painting, and lead battery manufacturing recycling applications.
  • Maintenance-free disposable hood
  • Easy-to-clean belt saves time and effort
  • Standard capacity lithium ion - rechargeable battery
The standard battery lasts from 5.5 to 13 hours, delivering more run time, less charging time and reduced downtime.
Delivery: Includes a premium suspension hood, length adjusting breathing tube, breathing tube cover, PARP unit, standard battery, easy clean belt, single station battery charger, high efficiency filter, filter cover, spark arrestor, airflow indicator and prefilter.