Order Entry
United States
Orders LinkContactUsLinkComponent
 

DMNPE-caged Luciferin

Marketsource
Supplier:   OZ Biosciences
DMNPE-caged Luciferin
Supplier:  OZ Biosciences
Marketsource
Certificates
DMNPE-caged Luciferin
DMNPE-caged Luciferin

3 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The cell permeable DMNPE-caged Luciferin is as a bioluminescent luciferase substrate used to measure intracellular functions since it readily crosses cell membranes. It is a D-luciferin ester analogue of luciferin.

  • High purity
  • Cell permeable: efficient luciferin delivery into intact cells even at neutral pH
  • Ideal for in vivo applications
  • Induces prolonged release of luciferin by the action of cells’ esterases
  • Allows to follow changes in gene expression in live cells
  • Suitable for in vitro experiments

Once the caged luciferin is inside the cells, active luciferin can be released by the action of endogenous intracellular esterases or by a flash of UV light. This implies:
1. A more efficient delivery of luciferin into intact living cells & improved sensitivity analysis of in vivo luciferase assays
2. A long-term measurement of luciferase activity since hydrolysis by esterases allows a continuous supply of active Luciferin
DMNPE-caged luciferin appears a good alternative to D-Luciferin classically used to measure luciferase activity in live cells. Indeed, the difficulty to deliver D-Luciferin into living cells renders in vivo quantification limited to a certain extent- for example, permeability issues at neurtral pH.

Shipped at room temperature.
Store at -20°C and protect from light.

Synonyms:  D-Luciferin 1-(4,5-dimethoxy-2-nitrophenyl)ethyl ester
  • CAS Number:223920-67-0
  • Formula:C₂₁H₁₉N₃O₇S₂
  • Molecular Weight:489.52 g/mol
  •  :Packaged under nitrogen.
  • Storage Temperature:Freezer
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Bottle, amber
    10 mg
    Log in for availability
    Log in for availability
    Each
    Each - $219.97
    $219.97
    Each
       $219.97
     

    Supplier Number: LC10000
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource
  • Bottle, amber
    25 mg
    Log in for availability
    Log in for availability
    Each
    Each - $420.40
    $420.40
    Each
       $420.40
     

    Supplier Number: LC25000
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource
  • Bottle, amber
    50 mg
    Log in for availability
    Log in for availability
    Each
    Each - $768.12
    $768.12
    Each
       $768.12
     

    Supplier Number: LC50000
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource