ContactUsLinkComponent
DuPont™ Tyvek® IsoClean® Hood with Attached Mask
DuPont™ Tyvek® IsoClean® Hood with Attached Mask
Catalog # 76188-370
Supplier:  DuPont
undefined
DuPont™ Tyvek® IsoClean® Hood with Attached Mask
Catalog # 76188-370
Supplier:  DuPont
Supplier Number:  IC669BWH0001000S
Specifications
  • Sterile:
    +
  • Garment application:
    Cleanroom
  • Color:
    White
  • Cat. No.:
    76188-370
  • W×L:
    295/8×183/8"
Specifications
About this item
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Hood with attached 7" blue mask
  • Pleated outer mask
  • Bound seams
  • Bound head opening
  • Ties with loops
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash.
These masks feature an attached blue mask that measures 7" wide. Pleated outer mask is made of polyethylene. Seams and head opening are bound and feature ties and loops for a secure fit.
Certifications: Sterile garments are gamma sterilized to SAL of 10-6.
Packaging: Packaged in an ISO Class 5 cleanroom. Individually packaged in an opaque bag and placed in double transparent poly liners.