Order Entry
United States
ContactUsLinkComponent
Prepared Tryptic Soy Broth Media, Mesa Laboratories
Prepared Tryptic Soy Broth Media, Mesa Laboratories
Catalog # 101384-692
Supplier:  Mesa Labs
CAS Number:  
Prepared Tryptic Soy Broth Media, Mesa Laboratories
Catalog # 101384-692
Supplier:  Mesa Labs
Supplier Number:  77-04-TSB-BP16
CAS Number:  

Specifications

  • Description:
    Tryptic Soy Broth w/Bromocresol Purple
  • Size:
    6.5 mL
  • Cat. No.:
    101384-692
  • Supplier no.:
    TSB-BP16

Specifications

About this item

Soybean Casein Digest Broth is the recommended media for use with Biological Indicators. Use of another media may detrimentally affect outgrowth of spores. TSB with Bromocresol purple pH indicator is specially formulated for use with our biological indicators.

Packaging: Packaged in Type I borosilicate glass tubes (16×100 mm) with PP screw-caps (white rubber liners).