Alumel Calibration Reference Material, PerkinElmer
Catalog # 97012-174
Supplier: PerkinElmer
CAS Number:
Specifications
- Description:Alumel Reference Material
- Size:3×2" wire
- Cat. No.:97012-174
- Supplier no.:09988015
Specifications
About this item
Magnetic transition temperature of 154.2 °C, 3 x 2" pieces of alumel wire.