Order Entry
United States
ContactUsLinkComponent
Anti-Ovalbumin IgG1 (Mouse) ELISA Kit, Cayman Chemical
Anti-Ovalbumin IgG1 (Mouse) ELISA Kit, Cayman Chemical
Catalog # 75816-812
CAS Number:  
undefined
Anti-Ovalbumin IgG1 (Mouse) ELISA Kit, Cayman Chemical
Catalog # 75816-812
Supplier Number:  500830-96
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Size:
    96 Wells
  • Detection Range:
    1.56 – 200 ng/mL
  • Cat. No.:
    75816-812
  • Contents:
    Antibody, Standard, Buffer, Polysorbate 20, Substrate solution, Stop solution, Plate, and Cover Sheet
  • Supplier no.:
    500830-96

Specifications

About this item

For the measurement of anti-ovalbumin of the IgG1 isotype in mouse plasma and serum.

  • Measure mouse anti-OVA IgG1 in mouse plasma, serum, or BAL fluids
  • Employs an affinity-purified mouse anti-OVA IgG1 polyclonal antibody standard for accurate results
  • Assay Range: 1.56 – 200 ng/mL with an LLOQ of 1.56 ng/mL
  • Rapid assay; get results in under 4 hours

Cayman's Ovalbumin IgG1 (mouse) ELISA Kit is an immunometric (i.e., “sandwich”) assay which can be used to measure anti-ovalbumin of the IgG1 isotype in mouse plasma and serum without prior sample purification.