Slide/Cassette Drawer Storage System, Springside Scientific
Catalog # 76202-570
Supplier: SPRINGSIDE SCIENTIFIC, LLC.
Specifications
- Description:Slide/Cassette Drawer Storage System
- Dimensions:12W×11.5D×4" H
- Cat. No.:76202-570
Specifications
About this item
Economical plastic-coated cardboard Slide/Cassette Drawer Storage System.
Economical drawer system made of high-quality cardboard is designed for cassette block and slide storage. Made from plastic-coated cardboard. System includes 10 sets per package with 8 drawers available for each set, along with spacer, index tags, and labels.