About this item
Myogenin is a member of the MyoD family of myogenic basic helix-loop-helix (bHLH) transcription factors that also includes MyoD, Myf-5, and MRF4 (also known as herculinor Myf-6). MyoD family members are expressed exclusively in skeletal muscle and play a key role in activating myogenesis by binding to enhancer sequences of muscle-specific genes. The regulatory domain of MyoD is approximately 70 amino acids in length and includes both a basic DNA binding motif and a bHLH dimerization motif. MyoD family members share about 80% amino acid homology in their bHLH motifs. Anti-myogenin labels the nuclei of myoblasts in developing muscle tissue, and is expressed in tumor cell nuclei of rhabdomyosarcoma and some leiomyosarcomas. Positive nuclear staining may occur in Wilms tumor.
CF® dyes are Biotium's next-generation fluorescent dyes. CF®405S is a blue fluorescent dye (Ex/Em 404/431 nm) with superior brightness compared to other blue dyes; it is also compatible with super-resolution imaging by SIM. Note: Conjugates of blue fluorescent dyes are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.
Type: Primary
Antigen: MYOG
Clonality: Monoclonal
Clone: Clone MGN185 F5D
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Cat, Human, Mouse, Pig, Rat
Specifications
- Catalog No:
- 75965-036
- 75965-038
- Antigen Symbol:
- MYOG
- MYOG
- Antigen Name:
- Myogenin
- Myogenin
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 405S
- CF® 405S
- Clone:
- MGN185 F5D
- MGN185 F5D
- Reactivity:
- Pig, Human, Rat, Mouse, Cat
- Pig, Human, Rat, Mouse, Cat
- Host:
- Mouse
- Mouse
- Gene ID:
- Unigene ID: 2830
- Unigene ID: 2830
- Isotype:
- IgGs
- IgGs
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0.1 mg/mL
- 0.1 mg/mL
- Storage Buffer:
- PBS/0.1% BSA/0.05% azide
- PBS/0.1% BSA/0.05% azide