Order Entry
United States
ContactUsLinkComponent
Anti-KRT76 Mouse Monoclonal Antibody (CF405S) [clone: KRTH/1076]
Anti-KRT76 Mouse Monoclonal Antibody (CF405S) [clone: KRTH/1076]
Supplier:  Biotium
Certificates
Anti-KRT76 Mouse Monoclonal Antibody (CF405S) [clone: KRTH/1076]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75918-192
  • 75916-868
  • Antigen Symbol:
  • KRT76
  • KRT76
  • Antigen Name:
  • Cytokeratin HMW
  • Cytokeratin HMW
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • KRTH/1076
  • KRTH/1076
  • Reactivity:
  • Human, Rat
  • Human, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 654392
  • Unigene ID: 654392
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications

Frequently Bought Together