Order Entry
United States
ContactUsLinkComponent
Anti-NKX2-1 Mouse Monoclonal Antibody (CF594) [clone: 8G7G3/1 NX2.1/690]
Anti-NKX2-1 Mouse Monoclonal Antibody (CF594) [clone: 8G7G3/1 NX2.1/690]
Supplier:  Biotium
Certificates
Anti-NKX2-1 Mouse Monoclonal Antibody (CF594) [clone: 8G7G3/1 NX2.1/690]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75978-190
  • 75978-188
  • Antigen Symbol:
  • NKX2-1
  • NKX2-1
  • Antigen Name:
  • TTF-1 / NKX2.1
  • TTF-1 / NKX2.1
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • 8G7G3/1 NX2.1/690
  • 8G7G3/1 NX2.1/690
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 94367
  • Unigene ID: 94367
  • Isotype:
  • IgG
  • IgG
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications

Frequently Bought Together