Order Entry
United States
ContactUsLinkComponent
Anti-DPP4 Mouse Monoclonal Antibody [clone: 202-36]
Anti-DPP4 Mouse Monoclonal Antibody [clone: 202-36]
Supplier:  Biotium
Certificates
Anti-DPP4 Mouse Monoclonal Antibody [clone: 202-36]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75890-992
  • 75890-994
  • Antigen Symbol:
  • DPP4
  • DPP4
  • Antigen Name:
  • CD26
  • CD26
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 202-36
  • 202-36
  • Reactivity:
  • Human, Rat
  • Human, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 368912
  • Unigene ID: 368912
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.2 mg/mL
  • 0.2 mg/mL
  • Storage Buffer:
  • PBS/0.05% BSA/0.05% azide
  • PBS/0.05% BSA/0.05% azide

Specifications

Frequently Bought Together