Order Entry
United States
ContactUsLinkComponent
Anti-KRT10 Mouse Monoclonal Antibody [clone: LH2]
Anti-KRT10 Mouse Monoclonal Antibody [clone: LH2]
Supplier:  Biotium
Certificates
Anti-KRT10 Mouse Monoclonal Antibody [clone: LH2]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75916-100
  • 75916-098
  • Antigen Symbol:
  • KRT10
  • KRT10
  • Antigen Name:
  • Cytokeratin 10
  • Cytokeratin 10
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • LH2
  • LH2
  • Reactivity:
  • Human, Mouse
  • Human, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 99936
  • Unigene ID: 99936
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.2 mg/mL
  • 0.2 mg/mL
  • Storage Buffer:
  • PBS/0.05% BSA/0.05% azide
  • PBS/0.05% BSA/0.05% azide

Specifications

Frequently Bought Together