Order Entry
United States
ContactUsLinkComponent
Anti-MYOG Mouse Monoclonal Antibody (CF488A) [clone: F5D]
Anti-MYOG Mouse Monoclonal Antibody (CF488A) [clone: F5D]
Supplier:  Biotium
Certificates
Anti-MYOG Mouse Monoclonal Antibody (CF488A) [clone: F5D]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75964-880
  • 75964-914
  • Antigen Symbol:
  • MYOG
  • MYOG
  • Antigen Name:
  • Myogenin
  • Myogenin
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • F5D
  • F5D
  • Reactivity:
  • Pig, Human, Dog, Rat, Mouse, Cat
  • Pig, Human, Dog, Rat, Mouse, Cat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 2830
  • Unigene ID: 2830
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications

Frequently Bought Together