About this item
This antibody recognizes a protein of 205 kDa, which is identified as integrin beta-4 (ITGB4). Its epitope is localized in the extracellular domain of ITGB4 protein. Integrins are heterodimers comprised of alpha and beta subunits, that are non-covalently associated transmembrane glycoprotein receptors. Different combinations of alpha and beta polypeptides form complexes that vary in their ligand-binding specificities. Integrins mediate cell-matrix or cell-cell adhesion, and transduced signals that regulate gene expression and cell growth. This gene encodes the integrin beta-4 subunit, a receptor for the laminins. This subunit tends to associate with alpha-6 subunit and is likely to play a pivotal role in the biology of invasive carcinoma. Mutations in this gene are associated with epidermolysis bullosa with pyloric atresia. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Type: Primary
Antigen: ITGB4
Clonality: Monoclonal
Clone: Clone UM-A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 75883-360
- 75883-358
- Antigen Symbol:
- ITGB4
- ITGB4
- Antigen Name:
- CD104
- CD104
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- UM-A9
- UM-A9
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- Unigene ID: 632226
- Unigene ID: 632226
- Isotype:
- IgG2a kappa
- IgG2a kappa
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0.2 mg/mL
- 0.2 mg/mL
- Storage Buffer:
- PBS/0.05% BSA/0.05% azide
- PBS/0.05% BSA/0.05% azide