Order Entry
United States
ContactUsLinkComponent
Anti-MS4A1 Mouse Monoclonal Antibody (CF594) [clone: B9E9]
Anti-MS4A1 Mouse Monoclonal Antibody (CF594) [clone: B9E9]
Supplier:  Biotium
Certificates
Anti-MS4A1 Mouse Monoclonal Antibody (CF594) [clone: B9E9]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75890-622
  • 75890-624
  • Antigen Symbol:
  • MS4A1
  • MS4A1
  • Antigen Name:
  • CD20
  • CD20
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 594
  • CF® 594
  • Clone:
  • B9E9
  • B9E9
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 712553
  • Unigene ID: 712553
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications

Frequently Bought Together