Order Entry
United States
ContactUsLinkComponent
 
Anti-POMC Mouse Monoclonal Antibody (CF488A) [clone: 57]
 :  Biotium
 
Anti-POMC Mouse Monoclonal Antibody (CF488A) [clone: 57]
 :  Biotium

 

  • Catalog No:
  • 75883-644
  • 75883-642
  • Antigen Symbol:
  • POMC
  • POMC
  • Antigen Name:
  • ACTH
  • ACTH
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • 57
  • 57
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 1897
  • Unigene ID: 1897
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

 

Frequently Bought Together