Order Entry
United States
ContactUsLinkComponent
Anti-p55 Mouse Monoclonal Antibody (CF405S) [clone: 1108-1]
Anti-p55 Mouse Monoclonal Antibody (CF405S) [clone: 1108-1]
Supplier:  Biotium
Certificates
Anti-p55 Mouse Monoclonal Antibody (CF405S) [clone: 1108-1]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75967-748
  • 75967-750
  • Antigen Symbol:
  • p55
  • p55
  • Antigen Name:
  • 50 EBV-Early Antigen
  • 50 EBV-Early Antigen
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • 1108-1
  • 1108-1
  • Reactivity:
  • EBV (Epstein-Barr virus)
  • EBV (Epstein-Barr virus)
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications

Frequently Bought Together