Order Entry
United States
ContactUsLinkComponent
Anti-HIF1A Mouse Monoclonal Antibody (CF405S) [clone: HIF1A-84]
Anti-HIF1A Mouse Monoclonal Antibody (CF405S) [clone: HIF1A-84]
Supplier:  Biotium
Certificates
Anti-HIF1A Mouse Monoclonal Antibody (CF405S) [clone: HIF1A-84]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75952-824
  • 75952-826
  • Antigen Symbol:
  • HIF1A
  • HIF1A
  • Antigen Name:
  • HIF1-alpha
  • HIF1-alpha
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • HIF1A-84
  • HIF1A-84
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 597216
  • Unigene ID: 597216
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications

Frequently Bought Together