Order Entry
United States
ContactUsLinkComponent
Anti-KRT18 Mouse Monoclonal Antibody (CF405S) [clone: C-04]
Anti-KRT18 Mouse Monoclonal Antibody (CF405S) [clone: C-04]
Supplier:  Biotium
Certificates
Anti-KRT18 Mouse Monoclonal Antibody (CF405S) [clone: C-04]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75918-392
  • 75918-390
  • Antigen Symbol:
  • KRT18
  • KRT18
  • Antigen Name:
  • Cytokeratin 18
  • Cytokeratin 18
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • C-04
  • C-04
  • Reactivity:
  • Rat, Mouse, Cat, Marmoset, Hamster, Pig, Horse, Monkey, Sheep, Goat, Ferret, Human, Bovine, Dog
  • Rat, Mouse, Cat, Marmoset, Hamster, Pig, Horse, Monkey, Sheep, Goat, Ferret, Human, Bovine, Dog
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 406013
  • Unigene ID: 406013
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications

Frequently Bought Together