Order Entry
United States
ContactUsLinkComponent
 
Human Factor Ⅺ Total Antigen Assay ELISA Kit, Molecular Inovations
 
Human Factor Ⅺ Total Antigen Assay ELISA Kit, Molecular Inovations

 

  • Conjugate ELISA:
    HRP-Streptavidin
  • Format:
    Pre-coated
  • Host:
    Human
  • Primary antibody reactivity:
    Human
  • Target protein:
    Factor XI
  • Detection Method:
    Photometric (450 nm)
  • Detection Range:
    0.2 - 100 ng/ml
  • Storage Temperature:
    4 °C

 

Frequently Bought Together