Order Entry
United States
ContactUsLinkComponent
 
ImageQuant™ Chemiluminescent Imaging System, LAS 500, Cytiva
Catalog # 102985-852
Supplier:  Avantor
undefined
ImageQuant™ Chemiluminescent Imaging System, LAS 500, Cytiva
Catalog # 102985-852
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Resolution:
    8.3 megapixel CCD
  • Description:
    ImageQuant LAS 500
  • Environmentally Preferable:
  • W×D×H:
    300×280×570 mm
  • Cat. No.:
    102985-852
  • Supplier no.:
    29-0050-63

Specifications

About this item

ImageQuant™ LAS 500 is a cooled CCD imager for imaging of chemiluminescent Western blots, fluorescent protein and DNA gel stains, and white light imaging of colorimetric stains and markers.

  • Sensitivity: Detect picogram levels of sample with chemiluminescence
  • Automatic overlay function: Simultaneously image chemiluminescent samples and colored molecular weight markers
  • Ready-to-operate in less than 5 min with the rapid cooling camera
  • Compact integrated design works independently of a dedicated PC with flexible image storage
  • Flexible storage of image on a USB memory stick, on the system or in a network connection folder
  • 4.8 orders of magnitude

ImageQuant™ LAS 500 is a practical and affordable solution to individual researchers or laboratories that are either looking for their own imager or need extra imaging capacity. The 300×280 mm footprint has the same area as a typical laptop computer, which allows the imager to fit neatly on almost any lab bench without the need to connect ancillary equipment.

The CCD camera is ready for use in minutes, and advanced Peltier cooling and binning significantly reduce noise levels, increasing both the sensitivity and linearity of the system. It is designed for chemiluminescence detection with regard to sensitivity and dynamic range.

In addition, the system has three LED light sources; a combined blue epi-/UV, epi-light and a white epi-light. The combined blue epi-/UV and epi-light allows imaging of fluorescence signals above 560 nm. This feature makes the camera well suited for DNA stain imaging with SYBR™ Green and ethidium bromide (EtBr) as well as with common total protein stain dyes. The white epi-light source (470 to 635 nm) is for documentation of colorimetric markers and stains, such as Coomassie™ Blue or silver.

Delivery: Supplied with touchscreen, sample chamber, USB port, camera and light sources.