Specifications
- Method / Format:Magnetic beads
- Prep size:Mini
- Refine target molecule:poly(A) RNA
- Sample type:RNA preps
- Target molecule:RNA
- Description:PolyATtract mRNA Isolation System III with Magnetic Stand, 15 isolations
- Size:15 isolations
- Storage Conditions:Store at 4 °C. Do not freeze the MagneSphere Paramagnetic Particles.
- Description:PolyATtract mRNA Isolation System III with Magnetic Stand, 15 isolations
- Cat. No.:PAZ5300
- Supplier no.:Z5300
Specifications
About this item
The PolyATtract System isolates messenger RNA directly from total RNA.
- Isolates Highly Pure mRNA from Total RNA
- Protocol complete in approximately 45 minutes
- Isolated mRNA is suitable for use in in vitro translation, RT-PCR and cDNA synthesis
- Cat.# Z5200 and Z5210 contain sufficient reagents for 3 separate mRNA isolations, each from 1-5mg total RNA
- Cat.# Z5300 and Z5310 contain sufficient reagents for 15 separate mRNA isolations, each from 100-1,000µg total RNA
The PolyATtract mRNA Isolation Systems use the MagneSphere technology to isolate mRNA rapidly and effectively from total RNA. The systems use a biotinylated oligo(dT) primer to hybridize, at high efficiency in solution, to the 3' poly(A)+ region present in most mature eukaryotic mRNAs. The hybrids are bound to streptavidin coupled to paramagnetic particles, captured using a magnetic separation stand and washed at high stringency. The mRNA is eluted from the solid phase by the simple addition of ribonuclease-free, deionized water. With total RNA as the starting material, poly(A)+ mRNA is isolated in approximately 45 minutes. The isolated mRNA is suitable for all molecular biology applications, including in vitro translation and cDNA synthesis. Cat.# Z5200 contains sufficient reagents for 3 separate mRNA isolations, each from 1-5mg of total RNA. Cat.# Z5210 contains the same reagents as Cat.# Z5200, excluding the Magnetic Separation Stand. Cat.# Z5300 contains sufficient reagents for 15 separate mRNA isolations, each from 100-1,000ug of total RNA. Cat.# Z5310 contains the same reagents as Cat.# Z5300, excluding the Magnetic Separation Stand.