Order Entry
United States
ContactUsLinkComponent
PolyATtract mRNA Isolation System II with Magnetic Stand, 3 isolations, Promega
PolyATtract mRNA Isolation System II with Magnetic Stand, 3 isolations, Promega
Catalog # PAZ5200
CAS Number:  
undefined
PolyATtract mRNA Isolation System II with Magnetic Stand, 3 isolations, Promega
Catalog # PAZ5200
Supplier Number:  Z5200
CAS Number:  

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    poly(A) RNA
  • Sample type:
    RNA preps
  • Target molecule:
    RNA
  • Description:
    PolyATtract mRNA Isolation System II with Magnetic Stand, 3 isolations
  • Size:
    3 isolations
  • Storage Conditions:
    Store at 4 °C. Do not freeze the MagneSphere Paramagnetic Particles.
  • Description:
    PolyATtract mRNA Isolation System II with Magnetic Stand, 3 isolations
  • Cat. No.:
    PAZ5200
  • Supplier no.:
    Z5200

Specifications

About this item

The PolyATtract System isolates messenger RNA directly from total RNA.

  • Isolates Highly Pure mRNA from Total RNA
  • Protocol complete in approximately 45 minutes
  • Isolated mRNA is suitable for use in in vitro translation, RT-PCR and cDNA synthesis
  • Cat.# Z5200 and Z5210 contain sufficient reagents for 3 separate mRNA isolations, each from 1-5mg total RNA
  • Cat.# Z5300 and Z5310 contain sufficient reagents for 15 separate mRNA isolations, each from 100-1,000µg total RNA

The PolyATtract mRNA Isolation Systems use the MagneSphere technology to isolate mRNA rapidly and effectively from total RNA. The systems use a biotinylated oligo(dT) primer to hybridize, at high efficiency in solution, to the 3' poly(A)+ region present in most mature eukaryotic mRNAs. The hybrids are bound to streptavidin coupled to paramagnetic particles, captured using a magnetic separation stand and washed at high stringency. The mRNA is eluted from the solid phase by the simple addition of ribonuclease-free, deionized water. With total RNA as the starting material, poly(A)+ mRNA is isolated in approximately 45 minutes. The isolated mRNA is suitable for all molecular biology applications, including in vitro translation and cDNA synthesis. Cat.# Z5200 contains sufficient reagents for 3 separate mRNA isolations, each from 1-5mg of total RNA. Cat.# Z5210 contains the same reagents as Cat.# Z5200, excluding the Magnetic Separation Stand. Cat.# Z5300 contains sufficient reagents for 15 separate mRNA isolations, each from 100-1,000ug of total RNA. Cat.# Z5310 contains the same reagents as Cat.# Z5300, excluding the Magnetic Separation Stand.