Order Entry
United States
ContactUsLinkComponent
ADP-Glo Kinase Assay + TNIK Kinase Enzyme System, 1 each, Promega
ADP-Glo Kinase Assay + TNIK Kinase Enzyme System, 1 each, Promega
Catalog # PAV4159
CAS Number:  
undefined
ADP-Glo Kinase Assay + TNIK Kinase Enzyme System, 1 each, Promega
Catalog # PAV4159
Supplier Number:  V4159
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    ADP-Glo Kinase Assay + TNIK Kinase Enzyme System, 1 each
  • Size:
    1 each
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAV4159
  • Supplier no.:
    V4159

Specifications

About this item

Recombinant human TNIK (1-367) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. TNIK or TRAF2 and NCK-interacting kinase are characterized by N-terminal kinase and C-terminal GCK domains that serve a regulatory function.

  • Easily Screen and Profile TNIK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Recombinant human TNIK (1-367) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. TNIK or TRAF2 and NCK-interacting kinase are characterized by an N-terminal kinase domain and a C-terminal GCK domain that serve a regulatory function (1). TNIK is mainly expression in brain, heart and spleen and is a specific effector of RAP2, which regulates actin cytoskeleton (2). TNIK is autophosphorylated in a manner dependent upon Lys54 in the ATP-binding pocket of its kinase domain and plays a main role in cytoskeleton regulation. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: TNIK, 10ug (Human, recombinant; amino acids 1-367). MW: ~67kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT, MnCl2. TNIK NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/23043/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.