Order Entry
United States
ContactUsLinkComponent
ADP-Glo Kinase Assay + RSK1 Kinase Enzyme System, 1 each, Promega
ADP-Glo Kinase Assay + RSK1 Kinase Enzyme System, 1 each, Promega
Catalog # PAV4047
CAS Number:  
undefined
ADP-Glo Kinase Assay + RSK1 Kinase Enzyme System, 1 each, Promega
Catalog # PAV4047
Supplier Number:  V4047
CAS Number:  

Specifications

  • Description:
    ADP-Glo Kinase Assay + RSK1 Kinase Enzyme System, 1 each
  • Size:
    1 each
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAV4047
  • Supplier no.:
    V4047

Specifications

About this item

Recombinant full-length human RSK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. RSK1 is a member of the RSK (ribosomal S6 kinase) family that consists of growth factor-regulated serine/threonine kinases.

  • Easily Screen and Profile RSK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Recombinant full-length human RSK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. RSK1 is a member of the RSK (ribosomal S6 kinase) family that consists of growth factor-regulated serine/threonine kinases. RSK1 contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signaling pathway. RSK1 encodes a predicted 735-amino acid protein containing 2 distinct consensus ATP-binding site sequences. RSK1 transcript is present in lymphocytes, skeletal muscle, liver and adipose tissue (1). RSKs are implicated in the activation of the mitogen-activated kinase (MAPK) cascade and the stimulation of cell proliferation and differentiation (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: RSK1, 10ug (Human, recombinant full-length). MW: ~108kDa. Substrate: S6K Substrate (KRRRLASLR); derived from human 40S ribosomal protein S6 (amino acids 230-238). Other: Reaction Buffer, DTT. RSK1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/6195/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.