Specifications
- Description:ADP-Glo Kinase Assay + RET (V804L) Kinase Enzyme System, 1 each
- Size:1 each
- Storage Temperature:°COMPONENTS AT DIFFERENT TEMPS
- Cat. No.:PAV4473
- Supplier no.:V4473
Specifications
About this item
Recombinant human RET (V804L) (amino acids 658-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
- Easily Screen and Profile RET (V804L) Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human RET (V804L) (amino acids 658-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. RET gene codes for a transmembrane tyrosine kinase, which is a subunit of a multimeric complex that acts as a receptor for four structurally related molecules: GDNF, neurturin, artemin and persephin. Germline mutations of RET cause a dominantly inherited dysgenesis of the enteric nervous system known as Hirschsprung's disease. RET is constitutively activated by point mutations in hereditary medullary thyroid carcinomas (MTCs). Several single nucleotide polymorphisms of the RET gene have been described. Multiple endocrine neoplasia type 2A (MEN 2A) have been reported to be associated with two mutations of the proto-oncogene RET. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: RET (V804L), 10microg (Human, recombinant; amino acids 658-end). MW: ~74kDa. Substrate: IGF1Rtide (KKKSPGEYVNIEFG); derived from human IRS-1 protein residues 891-902. Other: Reaction Buffer, DTT. RET (V804L) NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/5979/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.