Order Entry
United States
ContactUsLinkComponent
ADP-Glo Kinase Assay + PKC beta I Kinase Enzyme System, 1 each, Promega
ADP-Glo Kinase Assay + PKC beta I Kinase Enzyme System, 1 each, Promega
Catalog # PAV5095
CAS Number:  
undefined
ADP-Glo Kinase Assay + PKC beta I Kinase Enzyme System, 1 each, Promega
Catalog # PAV5095
Supplier Number:  V5095
CAS Number:  

Specifications

  • Description:
    ADP-Glo Kinase Assay + PKC beta I Kinase Enzyme System, 1 each
  • Size:
    1 each
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAV5095
  • Supplier no.:
    V5095

Specifications

About this item

Recombinant full-length human PKCbeta I was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PKCβI is a member of the PKC family (phospholipid-dependent serine/threonine kinase) and is highly related to PKCβ II.

  • Easily Screen and Profile PKCβI Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Recombinant full-length human PKCbeta I was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PKCβI is a member of the PKC family (phospholipid-dependent serine/threonine kinase) and is highly related to PKCβ II. Unlike the mature PKCβ II mRNA and protein, which rapidly increase following acute insulin treatment, the PKCβI mRNA and protein levels remain unchanged. The stable overexpression of PKCβI, but not PKCβI, leads to insulin-stimulated glucose uptake into cells. Upon stimulation of B lymphocytes and mast cells, Syk regulates Btk, and Btk selectively regulates enzymatic activity of PKCβI. Specific regulation of PKCβI by Btk is consistent with the selective association of Btk with PKCβI. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: PKCβI, 10microg (Human, recombinant full length). MW: ~102kDa. Substrate: PKCtide (ERMRPRKRQGSVRRRV); derived from protein kinase C epsilon (amino acid 149-164). Other: Reaction Buffer, DTT, Lipid Activator Solution. PKCβI NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/5579/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.