- Description:ADP-Glo Kinase Assay + PASK Kinase Enzyme System, 1 each
- Size:1 each
- Storage Temperature:°COMPONENTS AT DIFFERENT TEMPS
- Cat. No.:PAV4241
- Supplier no.:V4241
Recombinant human PASK (981-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PASK or PAS domain containing serine/threonine kinase regulates the function of many intracellular signaling pathways involved in stress.
- Easily Screen and Profile PASK Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human PASK (981-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PASK or PAS domain containing serine/threonine kinase regulates the function of many intracellular signaling pathways involved in stress. PASK is involved in sensing environmental changes in light intensity, oxygen concentration and redox potential. Through interaction with IRS-1, PASK has been proposed as a counter-regulatory mechanism in insulin and cytokine signaling (1). PASK can phosphorylate and inactivate glycogen synthase in vitro. Efficient phosphorylation requires residues 444 to 955 of PASK between the PAS and catalytic kinase domain, and this interaction is inhibited by the PAS domain. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: PASK, 10ug (Human, recombinant; amino acids 981-end). MW: ~66kDa. Substrate: ZIPtide (KKLNRTLSFAEPG). Other: Reaction Buffer, DTT. PASK NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/23178/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.