Order Entry
United States
ContactUsLinkComponent
ADP-Glo Kinase Assay + p70S6Kb Kinase Enzyme System, 1 each, Promega
ADP-Glo Kinase Assay + p70S6Kb Kinase Enzyme System, 1 each, Promega
Catalog # PAV4031
CAS Number:  
undefined
ADP-Glo Kinase Assay + p70S6Kb Kinase Enzyme System, 1 each, Promega
Catalog # PAV4031
Supplier Number:  V4031
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    ADP-Glo Kinase Assay + p70S6Kb Kinase Enzyme System, 1 each
  • Size:
    1 each
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAV4031
  • Supplier no.:
    V4031

Specifications

About this item

Recombinant full-length human p70S6Kb was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. p70S6Kb is a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases and is activated by mitogenic stimuli.

  • Easily Screen and Profile p70S6Kb Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Recombinant full-length human p70S6Kb was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. p70S6Kb is a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases and is activated by mitogenic stimuli, including growth factors, cytokines and phorbol esters. p70S6Kb contains 2 nonidentical kinase catalytic domains and phosphorylates the S6 ribosomal protein and eukaryotic translation initiation factor 4B. Phosphorylation of S6 leads to an increase in protein synthesis and cell proliferation. PI3 kinase pathway and mTOR are involved in the activation of p70S6Kb, but other pathways can also activate this target protein. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: p70S6Kb, 10ug (Human, recombinant full-length). MW: ~85kDa. Substrate: RSK substrate (KRRRLSSLRA); derived from human 40S ribosomal protein S6 (amino acids 230-239). Other: Kinase Assay Buffer I, DTT. p70S6Kb NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/6199/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.