Order Entry
United States
ContactUsLinkComponent
ADP-Glo Kinase Assay + NEK3 Kinase Enzyme System, 1 each, Promega
ADP-Glo Kinase Assay + NEK3 Kinase Enzyme System, 1 each, Promega
Catalog # PAV4501
CAS Number:  
undefined
ADP-Glo Kinase Assay + NEK3 Kinase Enzyme System, 1 each, Promega
Catalog # PAV4501
Supplier Number:  V4501
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    ADP-Glo Kinase Assay + NEK3 Kinase Enzyme System, 1 each
  • Size:
    1 each
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAV4501
  • Supplier no.:
    V4501

Specifications

About this item

Recombinant full-length human NEK3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. NEK3 is a member of the NEK family of protein kinases that share high amino acid homology with NIMA (never in mitosis gene a).

  • Easily Screen and Profile NEK3 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Recombinant full-length human NEK3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. NEK3 is a member of the NEK family of protein kinases that share high amino acid homology with NIMA (never in mitosis gene a). NEK3 mRNA is detected in all the proliferating cell lines with the amount not changing during the cell cycle. During Prolactin receptor signaling, VAV2 is phosphorylated and activated by NEK3. Overexpression of NEK3 in Chinese hamster ovary cells increases cytoskeletal re-organization in response to Prolactin while downregulation of NEK3 expression by siRNA blocks these effects. Prolactin also stimulates interaction between NEK3 and paxillin leading to increased paxillin phosphorylation. Analysis of breast tissue microarrays show a significant up-regulation of NEK3 expression in malignant versus normal specimens. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: NEK3, 10microg (Human,recombinant full length). MW: ~86kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT. NEK3 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/4752/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.