Order Entry
United States
ContactUsLinkComponent
 
Wizard Plus Minipreps DNA Purification System, Promega®
 
undefined
Wizard Plus Minipreps DNA Purification System, Promega®
 

 

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Plasmid
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Sample Size:
    1–3 mL of culture
  • Description:
    Wizard Plus Minipreps DNA Purification System
  • Yield:
    1–10 µg

 

 

Frequently Bought Together