Order Entry
United States
ContactUsLinkComponent
ADP-Glo Kinase Assay + DYRK2 Kinase Enzyme System, 1 each, Promega
ADP-Glo Kinase Assay + DYRK2 Kinase Enzyme System, 1 each, Promega
Catalog # PAV5091
CAS Number:  
undefined
ADP-Glo Kinase Assay + DYRK2 Kinase Enzyme System, 1 each, Promega
Catalog # PAV5091
Supplier Number:  V5091
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    ADP-Glo Kinase Assay + DYRK2 Kinase Enzyme System, 1 each
  • Size:
    1 each
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAV5091
  • Supplier no.:
    V5091

Specifications

About this item

Recombinant full-length human DYRK2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.

  • Easily Screen and Profile DYRK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Recombinant full-length human DYRK2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. DYRK2 (also known as dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2) belongs to a family of protein kinases whose members are presumed to be involved in cellular growth and/or development and is defined by structural similarity of their kinase domains and their capability to autophosphorylate on tyrosine residues. DYRK2 expressed in E. coli demonstrated tyrosine autophosphorylation and catalyzed phosphorylation of histones H3 and H2B in vitro. DYRK2 also regulates p53 to induce apoptosis in response to DNA damage. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: DYRK2, 10microg (Human, recombinant full-length). MW: ~95kDa. Substrate: DYRKtide (RRRFRPASPLRGPPK). Other: Reaction Buffer, DTT. DYRK2 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/8445/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.