Order Entry
United States
ContactUsLinkComponent
Wizard SV Gel and PCR Clean-Up Start-Up Kit, 110 V Electrical (500 Preps, Manifold and Free Vacuum Pump), 1 Each, Promega
Wizard SV Gel and PCR Clean-Up Start-Up Kit, 110 V Electrical (500 Preps, Manifold and Free Vacuum Pump), 1 Each, Promega
Catalog # PAA6750
CAS Number:  
Wizard SV Gel and PCR Clean-Up Start-Up Kit, 110 V Electrical (500 Preps, Manifold and Free Vacuum Pump), 1 Each, Promega
Catalog # PAA6750
Supplier Number:  A6750
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    DNA gel bands
    Amplified/modified DNA
  • Sample type:
    Gel matrix
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    Wizard SV Gel and PCR Clean-Up Start-Up Kit, 110V Electrical (500 preps, manifold and free vacuum pump), 1 each
  • Size:
    1 each
  • Environmentally Preferable:
  • Storage Conditions:
    15°C to 30°C
  • Description:
    Wizard SV Gel and PCR Clean-Up Start-Up Kit
  • Cat. No.:
    PAA6750
  • Supplier no.:
    A6750

Specifications

About this item

The Wizard SV Gel and PCR Clean-Up System is designed to purify DNA fragments of 100bp-10kb from standard or low-melting agarose gels or to purify products directly from PCR or restriction digests.

  • Bundle of 500 DNA Purification Kits, Vacuum Manifold and Free Vacuum Pump
  • Purify 100bp to 10kb DNA from Gel Slices or PCR
  • Centrifuge or vacuum-based protocols

The Wizard SV Gel and PCR Clean-Up System is designed to extract and purify DNA fragments of 100bp to 10kb from standard or low-melting agarose gels or to purify products directly from PCR and other common reactions such as restriction digests. Up to 95% recovery is achieved depending upon the DNA fragment size. PCR products are commonly purified to remove excess nucleotides and primers. This membrane-based system, which can bind up to 40ug of DNA, allows recovery of isolated DNA fragments or PCR products in as little as 15 minutes, depending on the number of samples processed. The purified DNA can be used for automated fluorescent DNA sequencing, cloning, labeling, restriction enzyme digestion or in vitro transcription/translation without further manipulation.