Order Entry
United States
Orders LinkContactUsLinkComponent
Proteinase K

Proteinase K

23 ml
Catalog # PAA5051
Supplier:   Promega Corporation
Supplier #:  A5051
Proteinase K
Catalog # PAA5051
CAS Number:  
Proteinase K
Proteinase K

23 ml

CAS Number:  
Price
$516.62
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Proteinase K, produced by the fungus Tritirachium album Limber, is a serine protease that exhibits broad cleavage activity. Proteinase K (PK) Solution is supplied at a concentration of 20 mg/ml in 10mM Tris-HCl (pH 7.5), 1 mM calcium chloride, 50% glycerol.

  • Stable at Room Temperature and Easy to Use
  • Active over pH range 4.3-12.0 in 0.5% SDS or 1% Triton(R) X-100
  • Retains >80% activity at temperatures up to 60°C
  • No resuspension or thawing before use
  • Cat.# MC5005, MC5008 supplied at a concentration of 20mg/ml

Proteinase K, produced by the fungus Tritirachium album Limber, is a serine protease that exhibits broad cleavage activity. It cleaves peptide bonds adjacent to the carboxylic group of aliphatic and aromatic amino acids and is useful for general digestion of protein in biological samples. It has been purified to remove RNase and DNase activities. The stability of Proteinase K in urea and SDS and its ability to digest native proteins make it useful for a variety of applications including preparation of chromosomal DNA for pulsed-field gel electrophoresis, protein fingerprinting and removal of nucleases from preparations of DNA and RNA. A typical working concentration for Proteinase K is 50-100ug/ml. Formulation: Proteinase K (PK) Solution is supplied at a concentration of 20mg/ml in 10mM Tris-HCl (pH 7.5), 1mM calcium chloride and 50% glycerol.

Documents

Specifications

Specifications

  • Size:
    23 ml
  • Storage Conditions:
    15...30 °C
  • Enzyme Synonyms:
    PK
  • Cat. No.:
    PAA5051
  • Enzyme Name:
    Proteinase K

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.