Order Entry
United States
ContactUsLinkComponent
ADP-Glo Kinase Assay + CAMKK1 Kinase Enzyme System, 1 each, Promega
ADP-Glo Kinase Assay + CAMKK1 Kinase Enzyme System, 1 each, Promega
Catalog # PAV4471
CAS Number:  
undefined
ADP-Glo Kinase Assay + CAMKK1 Kinase Enzyme System, 1 each, Promega
Catalog # PAV4471
Supplier Number:  V4471
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    ADP-Glo Kinase Assay + CAMKK1 Kinase Enzyme System, 1 each
  • Size:
    1 each
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAV4471
  • Supplier no.:
    V4471

Specifications

About this item

CAMKK1 or CAMKK alpha is a Ca2+/calmodulin-dependent protein kinase that activates CAM-kinases I and IV via phosphorylation of their Thr(177) and Thr(196) residues, respectively.

  • Easily Screen and Profile CAMKK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Recombinant full-length human CAMKK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CAMKK1 or CAMKK alpha is a Ca2+/calmodulin-dependent protein kinase that activates CAM-kinases I and IV via phosphorylation of their Thr(177) and Thr(196) residues, respectively. Recent studies have shown that the activity of CAMKK1 is decreased upon phosphorylation by cAMP-dependent protein kinase (PKA) (1). CAMKKI has been identified in intact cells as AMPKKs, predicting a significant role for this kinase in regulating AMPK activity in vivo. It has been shown that 2-deoxyglucose- and ionomycin-stimulated AMPK activity is substantially reduced in HeLa cells transfected with small interfering RNAs specific for CAMKKI (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: CAMKK1, 10ug (Human, recombinant full-length). MW: ~94kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT, Ca2+/Calmodulin Solution. CAMKK1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/84254/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.