To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Plasmid
- Sample type:Bacteria
- Target molecule:Plasmid
- Description:Wizard Plus SV Minipreps DNA Purification System + Vacuum Adapters
- Size:50 preps
- Environmentally Preferable:
- Sample Size:1–10 mL of culture
- Description:Wizard Plus SV Minipreps DNA Purification System
- Yield:1–20 µg
- Cat. No.:PAA1340
- Supplier no.:A1340
Specifications
About this item
A silica membrane-based system for simple, rapid isolation of plasmid DNA from 1-10ml E. coli cultures. The miniprep procedure can be completed in 45 minutes or less.
- 1-20microg High-Quality Plasmid DNA from 1-10ml Cultures
- Process up to 20 minipreps in 45 minutes
- Choice of spin or vacuum purification formats
- No phenol extractions or precipitations required
The Wizard Plus SV Minipreps DNA Purification System, a silica membrane-based system, provides a simple and reliable method for rapid isolation of plasmid DNA. The entire miniprep procedure can be completed in 45 minutes or less, depending on the number of samples processed. Using the system, plasmid DNA can be purified from 1-10ml of overnight E. coli culture. The purified plasmid DNA can be used directly for automated fluorescent BigDye terminator DNA sequencing as well as for other standard molecular biology techniques without further manipulation. It also can be used for in vitro transcription reactions when supplemented with a ribonuclease inhibitor such as Recombinant RNasin Ribonuclease Inhibitor (Cat.# N2511).