Order Entry
United States
ContactUsLinkComponent
1 kb DNA Step Ladder, 90 µg, Promega
1 kb DNA Step Ladder, 90 µg, Promega
Catalog # PAG6941
CAS Number:  
undefined
1 kb DNA Step Ladder, 90 µg, Promega
Catalog # PAG6941
Supplier Number:  G6941
CAS Number:  

Specifications

  • Description:
    1kb DNA Step Ladder, 90microg
  • Size:
    90 μg
  • Storage Temperature:
    -30°C to -10°C
  • Cat. No.:
    PAG6941
  • Supplier no.:
    G6941

Specifications

About this item

Ten blunt-ended DNA fragments ranging from 1kb to 10kb in 1kb increments.

  • Ten Blunt-Ended DNA Fragments of 1-10kb in 1kb Increments
  • Load 1microl/lane
  • Supplied at a concentration of 0.3µg/microl
  • Supplied with 6X Blue/Orange Loading Dye

The DNA Step Ladders are ladders of defined sizes with exact incremental steps between bands. The ladders are not intended for use in quantitative analysis. Each ladder is provided with a tube of 6X Blue/Orange Loading Dye. The fragments may be stained with ethidium bromide. 1kb DNA Step Ladder: Ten blunt-ended DNA fragments ranging from 1kb to 10kb in 1kb increments. All bands except the 5kb band are of equal intensity; the 5kb band is ~3 times more intense.