Order Entry
United States
ContactUsLinkComponent
Wizard Genomic DNA Purification Kit, Promega®
Wizard Genomic DNA Purification Kit, Promega®
Certificates
undefined
Wizard Genomic DNA Purification Kit, Promega®
Certificates

Specifications

  • Method / Format:
    Precipitation
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Plant samples
    Bacteria
    Biofluids (serum, plasma, saliva)
    Yeast/Fungi
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Sample Size:
    Scalable
  • Description:
    Wizard Genomic DNA Purification Kit
  • Yield:
    Varies based on starting material.

Specifications

Related Information

Frequently Bought Together