HYPE-CHO Transfection Kit, OZ Biosciences
Supplier: OZ Biosciences
Certificates
About this item
HYPE-CHO™ Transfection Kit has been designed for large scale up transient transfection and high protein expression such as antibody. The system is optimized for maximum efficiency in all CHO-S cells.
- Reach optimal Protein & Antibody production yield
- Suitable for all CHO cells growing in suspension
- Compatible for any synthetic or regular media used for protein production
- Ideal for bioreactor, spinner or flasks
- Animal origin free
It has been used and validated with cells from different origins (CHO, CHO-S, rCHO or any CHO-related cells) cultured in suspension (flask, spinner and bioreactor) used to produce proteins.
Delivery: Shipped at room temperature.
Packaging: Store Hype-CHO Reagent +4 °C - BCHO +4 °C
Specifications
- Bioproduction:Yes
- Cell Specific:CHO suspension